Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOX Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB1002527W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002527W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002527W Supplier Novus Biologicals Supplier No. NB1002527AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LOX Polyclonal specifically detects LOX in Human, Mouse, Pig, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown, Western Blot.

Specifications

Antigen LOX
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, KnockDown Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P28300
Gene Alias lysyl oxidaseEC 1.4.3.13, MGC105112, protein-lysine 6-oxidase
Gene Symbols LOX
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human LOX protein (within residues 200-300). [Swiss-Prot P28300].
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cytoskeleton Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4015
Test Specificity Human and mouse. Predicted to react with rat and pig based on 100% sequence homology. Immunogen sequence has 92% homology to cow and chicken and 85% homology to Xenopus.
Target Species Human, Mouse, Pig, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.