Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOX Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB1002530C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002530C 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002530C Supplier Novus Biologicals Supplier No. NB1002530C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LOX Polyclonal specifically detects LOX in Bovine, Human, Mouse, Pig, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen LOX
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P28300
Gene Alias lysyl oxidaseEC 1.4.3.13, MGC105112, protein-lysine 6-oxidase
Gene Symbols LOX
Host Species Rabbit
Immunogen A cocktail of two synthetic peptides; one made to a region of the human LOX protein within residues 300-350 (NB100-2528) and one within residues 200-300 (NB100-2527). [Swiss-Prot P28300]
Purification Method Immunogen affinity purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Cancer, Cytoskeleton Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4015
Test Specificity Human, mouse, rat, cow and pig. 92% sequence identity with chicken, Zebrafish, and Xenopus proteins.
Target Species Bovine, Human, Mouse, Pig, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.