Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOX Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB355062 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355062 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355062 Supplier Novus Biologicals Supplier No. NB1002527PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LOX Polyclonal specifically detects LOX in Human, Mouse, Pig, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown, Western Blot.

Specifications

Antigen LOX
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown, Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated
Formulation PBS
Gene Alias lysyl oxidaseEC 1.4.3.13, MGC105112, protein-lysine 6-oxidase
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human LOX protein (within residues 200-300). [Swiss-Prot P28300].
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4015
Target Species Human, Mouse, Pig, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.