Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOX propeptide Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. N1104168A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1104168A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1104168A94 Supplier Novus Biologicals Supplier No. NB11041568AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LOX propeptide Polyclonal antibody specifically detects LOX propeptide in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot.

Specifications

Antigen LOX propeptide
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias lysyl oxidaseEC 1.4.3.13, MGC105112, protein-lysine 6-oxidase
Gene Symbols LOX
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of mouse LOX propeptide (residues 78-115). [UniProtNo. P28301]
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Cytoskeleton Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4015.0
Test Specificity This antibody specifically recognizes both the pro-enzyme and pro-peptide forms of LOX protein in mouse and rat.
Target Species Human, Mouse, Rat, Pig, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.