Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LOXL3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$191.80 - $404.50
Specifications
Antigen | LOXL3 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEPA5145269
|
Invitrogen™
PA5145269 |
200 μL |
Each for $404.50
|
|
|||||
FEPA5145270
|
Invitrogen™
PA5145270 |
20 μL |
Each for $191.80
|
|
|||||
Description
LOXL3 is a member of the lysyl oxidase gene family. The prototypic member of the family is essential to the biogenesis of connective tissue, encoding an extracellular copper-dependent amine oxidase that catalyses the first step in the formation of crosslinks in collagens and elastin. A highly conserved amino acid sequence at the C-terminus end appears to be sufficient for amine oxidase activity, suggesting that each family member may retain this function. The N-terminus is poorly conserved and may impart additional roles in developmental regulation, senescence, tumor suppression, cell growth control, and chemotaxis to each member of the family.Specifications
LOXL3 | |
Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat, Pig | |
P58215, Q9Z175 | |
100512652, 16950, 312478, 84695 | |
Recombinant Human LOXL3 protein, Thr519-Asn726 (Accession #P58215). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
Lor2; LOW QUALITY PROTEIN: lysyl oxidase homolog 3; LOXL; Loxl2; Loxl3; Lysyl oxidase homolog 3; lysyl oxidase like 3; lysyl oxidase-like 2; lysyl oxidase-like 3; lysyl oxidase-like protein 3; Lysyl oxidase-related protein 2 | |
LOXL3 | |
IgG | |
Protein A | |
LOXL3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title