Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOXL4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP17425720 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17425720 20 μL
NBP174257 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP17425720 Supplier Novus Biologicals Supplier No. NBP17425720UL

Rabbit Polyclonal Antibody

LOXL4 Polyclonal specifically detects LOXL4 in Human samples. It is validated for Western Blot.

Specifications

Antigen LOXL4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q96JB6
Gene Alias EC 1.4.3, EC 1.4.3.-, FLJ21889, LOXClysyl oxidase related C, lysyl oxidase homolog 4, lysyl oxidase-like 4, Lysyl oxidase-like protein 4, Lysyl oxidase-related protein C
Gene Symbols LOXL4
Host Species Rabbit
Immunogen Synthetic peptides corresponding to the middle region of LOXL4. Immunizing peptide sequence ARGKLRPACPGGMHAVVSCVAGPHFRPPKTKPQRKGSWAEEPRVRLRSGA.
Molecular Weight of Antigen 82 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 84171
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.