Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LP 99
GREENER_CHOICE

Catalog No. p-7161649
Click to view available options
Quantity:
5 mg
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective BRD7 and BRD9 inhibitor (Kd values are 99 and 909 nM, respectively)

LP 99 is a selective BRD7 and BRD9 inhibitor (Kd values are 99 and 909 nM, respectively) that exhibits selectivity for BRD7/9 over a panel of 48 bromodomains (at 10 μM). LP 99 inhibits interactions with histones H3.3 and H4 at low micromolar concentrations, and decreases the level of IL-6 secreted from LPS-stimulated THP-1 cells, in vitro. A negative control for LP 99, ent-LP 99 (Cat.No. 5497), is also available.

Specifications

Chemical Name or Material N-[(2R,3S)-2-(4-Chlorophenyl)-1-(1,2-dihydro-1,4-dimethyl-2-oxo-7-quinolinyl)-6-oxo-3-piperidinyl]-2-methyl-1-propanesulfonamide
CAS 1808951-93-0
Quantity 25 mg
Molecular Formula C26H30ClN3O4S
Formula Weight 516.05
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.