Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LPAR1/LPA1/EDG-2 Antibody (1020714), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB345556 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345556 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345556 Supplier Novus Biologicals Supplier No. FAB99631MFV450

Mouse Monoclonal Antibody

LPAR1/LPA1/EDG-2 Monoclonal antibody specifically detects LPAR1/LPA1/EDG-2 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen LPAR1/LPA1/EDG-2
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 1020714
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias edg-2, EDG2Lysophosphatidic acid receptor Edg-2, endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor2,, Gpcr26, GPR26, LPA receptor 1, LPA-1, LPA1vzg-1, lysophosphatidic acid receptor 1, Mrec1.3, rec.1.3, ventricular zone gene 1, VZG1
Host Species Mouse
Immunogen E. coli-derived recombinant human LPAR1/LPA1/EDG-2, Met1-Lys50, Accession # Q92633
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1902
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.