Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LPAR1/LPA1/EDG-2 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP103363Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP103363Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP103363Z Supplier Novus Biologicals Supplier No. NBP103363AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LPAR1/LPA1/EDG-2 Polyclonal specifically detects LPAR1/LPA1/EDG-2 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen LPAR1/LPA1/EDG-2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias edg-2, EDG2Lysophosphatidic acid receptor Edg-2, endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor2,, Gpcr26, GPR26, LPA receptor 1, LPA-1, LPA1vzg-1, lysophosphatidic acid receptor 1, Mrec1.3, rec.1.3, ventricular zone gene 1, VZG1
Gene Symbols LPAR1
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human EDG2 (within residues 200-300). [Swiss-Prot# Q92633]
Molecular Weight of Antigen 43 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1902
Target Species Human, Rat, Chicken
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.