Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LPAR4/LPA4 Antibody (561203) [PerCP], Novus Biologicals™
SDP

Catalog No. NB402259 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB402259 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB402259 Supplier Novus Biologicals Supplier No. FAB10217C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LPAR4/LPA4 Monoclonal specifically detects LPAR4/LPA4 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen LPAR4/LPA4
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 561203
Conjugate PerCP
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias G protein-coupled receptor 23, GPR23P2Y purinoceptor 9, LPA4LPA-4, lysophosphatidic acid receptor 4, P2RY9G-protein coupled receptor 23, P2Y5-LIKE, P2Y5-like receptor, P2Y9LPA receptor 4, Purinergic receptor 9
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived transfected with human LPAR4/LPA4, Accession # Q99677
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2846
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.