Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LPAR5/LPA5/GPR92 Antibody (459812) [HRP], Novus Biologicals™
SDP

Catalog No. NB401951 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB401951 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB401951 Supplier Novus Biologicals Supplier No. FAB10297H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LPAR5/LPA5/GPR92 Monoclonal antibody specifically detects LPAR5/LPA5/GPR92 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF

Specifications

Antigen LPAR5/LPA5/GPR92
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 459812
Conjugate HRP
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready
Formulation PBS
Gene Alias G protein-coupled receptor 92, GPR92G-protein coupled receptor 92, GPR93G-protein coupled receptor 93, KPG_010, LPA receptor 5, LPA5LPA-5, lysophosphatidic acid receptor 5
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human LPAR5/LPA5/GPR92, Accession # Q9H1C0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 57121
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.