Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LPCAT2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | LPCAT2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
LPCAT2 Polyclonal specifically detects LPCAT2 in Mouse samples. It is validated for Western Blot.Specifications
LPCAT2 | |
Polyclonal | |
Rabbit | |
NP_766602 | |
54947 | |
The immunogen for this antibody is Lpcat2 - N-terminal region. Peptide sequence NENAQTPLVGRLLRALQPVLVSRVDPDSRKNTINEIKKRATSGGEWPQIL. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
1-alkylglycerophosphocholine O-acetyltransferase, Acetyl-CoA:lyso-PAF acetyltransferase, Acetyl-CoA:lyso-platelet-activating factor acetyltransferase, acyl-CoA:lysophosphatidylcholine acyltransferase 2, acyltransferase like 1, Acyltransferase-like 1, AYTL1LysoPAFAT, DKFZp686H22112, EC 2.3.1, EC 2.3.1.-, EC 2.3.1.23, EC 2.3.1.67,1-acylglycerophosphocholine O-acyltransferase, FLJ20481, LPC acyltransferase 2, LPCAT1, LPCAT-2, Lyso-PAF acetyltransferase, LysoPC acyltransferase 2, lysophosphatidylcholine acyltransferase 2, lyso-platelet-activating factor (PAF) acetyltransferase | |
LPCAT2 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title