Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LPPR3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP311908100UG
Description
LPPR3 Polyclonal antibody specifically detects LPPR3 in Human samples. It is validated for Western Blot, ELISASpecifications
LPPR3 | |
Polyclonal | |
Western Blot, ELISA | |
EC 3.1.3, EC 3.1.3.4, Lipid Phosphate Phosphatase-Related Protein Type 3, LPR3, PAP-2-Like Protein 2, PHP2, Plasticity-Related Gene 2 Protein, PRG2, PRG-2 | |
Synthetic peptide derived from the LPPR3 protein (Uniprot: Q6T4P5) | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
Unconjugated | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
79948 | |
Store at -70C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction