Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LPPR3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$219.50 - $487.50
Specifications
Antigen | LPPR3 |
---|---|
Dilution | Western Blot, ELISA |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
LPPR3 Polyclonal antibody specifically detects LPPR3 in Human samples. It is validated for Western Blot, ELISASpecifications
LPPR3 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Human | |
EC 3.1.3, EC 3.1.3.4, Lipid Phosphate Phosphatase-Related Protein Type 3, LPR3, PAP-2-Like Protein 2, PHP2, Plasticity-Related Gene 2 Protein, PRG2, PRG-2 | |
Synthetic peptide derived from the LPPR3 protein (Uniprot: Q6T4P5) | |
Primary | |
Store at -70C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
Polyclonal | |
Purified | |
RUO | |
PBS | |
79948 | |
IgG | |
Antigen Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title