Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ LPS from E. Coli, TLR4 ligand
SDP

Catalog No. p-200087446 Shop All R&D Systems Products
Click to view available options
Quantity:
1 mg

Applications: Functional

Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs). Use in functional reported in scientific literature (PMID 26121241).

Lipopolysaccharides from Escherichia coli 0111:B4 Gamma-irradiated, BioXtra, suitable for cell culture Dilution: Functional Purity: for ion exchange chromatography Reconstitute with 1 ml sterile balanced salt solution or tissue culture medium to the vial (1 mg) and gently swirl until the powder dissolves. Reconstituted product may be further diluted to desired working concentrations.

Specifications

Concentration LYOPH
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Quantity 1 mg
For Use With (Application) Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs). Use in functional reported in scientific literature (PMID 26121241).
Target LPS from E. Coli, TLR4 Ligand

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.