Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LpxT Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB287463 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287463 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287463 Supplier Novus Biologicals Supplier No. NBP323512AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LpxT Polyclonal antibody specifically detects LpxT in E. coli samples. It is validated for Western Blot, ELISA

Specifications

Antigen LpxT
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias ECK2168, Kdo(2)-lipid A phosphotransferase, Lipid A 1-diphosphate synthase, lipid A-core phosphotransferase, Undecaprenyl pyrophosphate:lipid A 1-phosphate phosphotransferase, yeiU
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid peptide near the carboxy terminus of E. coli LpxT. The immunogen is located within the last 50 amino acids of LpxT.
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 946693
Target Species E. coli
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.