Learn More
Invitrogen™ LR Clonase™ II Plus enzyme
Catalog No. 12538200
Encompass_Preferred
Click to view available options
Quantity:
100 Reactions
20 Reactions
Includes
Proteinase K solution (2μg/μL) and a positive control vector
Mix contains a proprietary blend of Int (Integrase), IHF (Integration Host Factor), and Xis (Excisionase) enzymes.
Gateway™ LR Clonase™ enzyme mix contains a proprietary blend of Int (Integrase), IHF (Integration Host Factor) and Xis (Excisionase) enzymes that catalyze the in vitro recombination between an Entry clone (containing a gene of interest flanked by attL sites) and a Destination vector (containing attR sites) to generate your expression clone. We offer different formats of LR Clonase enzyme mixtures, depending on your application and desired format.
LR Clonase II Plus enzyme mix is the latest version that offers the highest recombination efficiency available(Table 1), and is specifically optimized for both single and multiple fragment cloning (Table 1). LR Clonase II Plus enzyme mix is provided in an optimized single mix of enzyme and reaction buffer, ensuring enzyme stability and easy of use with few pippetting steps for both single and multiple fragment cloning. Our LR Clonase II enzyme mix is also available in a single mix format, while the original LR Clonase® enzyme is the enzyme and buffer in separate tubes.
Order Info
Shipping Conditions: Dry ice
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Includes LR Clonase II Plus enzyme and proteinase K solution (2 μg/μL). Store at -80°C. Guaranteed stable for 6 months when properly stored.
Shipping Condition Dry Ice
Enzyme LR Clonase
Compatible Buffer Reaction Buffer
Quantity 100 Reactions
Product Type LR Clonase Enzyme
Product Line Clonase™, Gateway™
WARNING: Cancer - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.