Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRAT Antibody (M34-P1F10), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB309395 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309395 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309395 Supplier Novus Biologicals Supplier No. NBP250444MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LRAT Monoclonal antibody specifically detects LRAT in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen LRAT
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone M34-P1F10
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias EC 2.3.1.135, LCA14, lecithin retinol acyltransferase, lecithin retinol acyltransferase (phosphatidylcholine--retinolO-acyltransferase), MGC33103, Phosphatidylcholine--retinol O-acyltransferase
Host Species Mouse
Immunogen Peptide Sequence - RDQRSVLASA (amino acids 190 -199)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Endocrinology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9227
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.