Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRBA Mouse anti-Human, Clone: 2D4.1, MilliporeSigma™
Mouse Monoclonal Antibody
$192.35 - $480.80
Specifications
Antigen | LRBA |
---|---|
Clone | 2D4.1 |
Concentration | Please refer to lot specific datasheet. |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF1096-25
|
MilliporeSigma
MABF109625UG |
25 μg |
Each for $192.35
|
|
|||||
MABF1096-100
|
MilliporeSigma
MABF1096100UG |
100 μg |
Each for $480.80
|
|
|||||
Description
Anti-LRBA, clone 2D4.1 Antibody.Specifications
LRBA | |
Please refer to lot specific datasheet. | |
Monoclonal | |
Purified | |
RUO | |
Human | |
LRBA,BGL,CDC4L,LBA | |
IgG1 κ | |
Protein G Purified | |
Clone 2D4.1 detects human Lipopolysaccharide-responsive and beige-like anchor protein (LRBA). It targets an epitope within 115 amino acids from the N-terminal half. |
2D4.1 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Inflammation and Immunology | |
NP_006717 | |
GST/His-tagged recombinant fragment corresponding to 115 amino acids from the N-terminal half of human Lipopolysaccharide-responsive and beige-like anchor protein (LRBA). | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title