Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRCH1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP244282 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP244282 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP244282 Supplier Novus Biologicals Supplier No. NBP244282
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects LRCH1 in Human samples, and it is validated for Immunoprecipitation, Western Blotting

LRCH1 Polyclonal specifically detects LRCH1 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen LRCH1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Alias CHDC1, FLJ41660, KIAA1016, leucine-rich repeat and calponin homology domain-containing protein 1, leucine-rich repeats and calponin homology (CH) domain containing 1, neuronal protein 81, NP81
Gene Symbols LRCH1
Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to a region between residue 375 to 425 of human Leucine-rich repeat and calponin homology domain-containing protein 1 using the numbering given in entry Q9Y2L9.3 (GeneID 23143).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23143
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.