Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRCH4 Antibody (OTI1F8), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N272520A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272520A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272520A405 Supplier Novus Biologicals Supplier No. NBP272520AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LRCH4 Monoclonal specifically detects LRCH4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen LRCH4
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1F8
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ40101, FLJ46315, leucine rich repeat neuronal 4, Leucine-rich neuronal protein, leucine-rich repeat and calponin homology domain-containing protein 4, Leucine-rich repeat neuronal protein 4, leucine-rich repeats and calponin homology (CH) domain containing 4, LRN, LRRN1, LRRN4, PP14183
Gene Symbols LRCH4
Host Species Mouse
Immunogen Full length recombinant protein of human LRCH4 (NP_002310) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4034.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.