Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRCH4 Antibody (OTI1F8), Novus Biologicals™
SDP

Catalog No. NBP245795 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP245795 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP245795 Supplier Novus Biologicals Supplier No. NBP245795
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot, Immunohistochemistry

LRCH4 Monoclonal specifically detects LRCH4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen LRCH4
Applications Immunohistochemistry (Paraffin), Immunohistochemistry
Classification Monoclonal
Clone OTI1F8
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150
Formulation PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
Gene Alias FLJ40101, FLJ46315, leucine rich repeat neuronal 4, Leucine-rich neuronal protein, leucine-rich repeat and calponin homology domain-containing protein 4, Leucine-rich repeat neuronal protein 4, leucine-rich repeats and calponin homology (CH) domain containing 4, LRN, LRRN1, LRRN4, PP14183
Gene Symbols LRCH4
Host Species Mouse
Immunogen Full length human recombinant protein of human LRCH4 (NP_002310) produced in HEK293T cell.
Molecular Weight of Antigen 73.3 kDa
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4034
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.