Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRDD Mouse anti-Mouse, DyLight 594, Clone: Lise-1, Novus Biologicals™
SDP

Catalog No. NB120607 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120607 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120607 Supplier Novus Biologicals Supplier No. NBP311674DL594

Mouse Monoclonal Antibody

LRDD Monoclonal specifically detects LRDD in Mouse samples. It is validated for Western Blot.

Specifications

Antigen LRDD
Applications Western Blot
Classification Monoclonal
Clone Lise-1
Conjugate DyLight 594
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias leucine-rich repeat and death domain-containing protein, leucine-rich repeats and death domain containing, MGC16925, p53-induced protein with a death domain, PIDD
Host Species Mouse
Immunogen Mouse LRDD (Death Domain).
Purification Method >95%, by SDS-PAGE
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55367
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.