Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRFN2 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB342621 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342621 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342621 Supplier Novus Biologicals Supplier No. NBP281731MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRFN2 Polyclonal antibody specifically detects LRFN2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen LRFN2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias fibronectin type III, immunoglobulin and leucine rich repeat domains 2, FIGLER2, leucine rich repeat and fibronectin type III domain containing 2, SALM1
Host Species Rabbit
Immunogen LRFN2 antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human LRFN2. The immunogen is located within amino acids 700 - 750 of LRFN2. Amino Acid Squence: PLEGKAKRSHSFD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 57497
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.