Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRFN5 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB356832 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB356832 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB356832 Supplier Novus Biologicals Supplier No. NBP177004MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRFN5 Polyclonal antibody specifically detects LRFN5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen LRFN5
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias C14orf146, chromosome 14 open reading frame 146, DKFZp686G0210, fibronectin type III, immunoglobulin and leucine rich repeat domains 8, FIGLER8, leucine rich repeat and fibronectin type III domain containing 5, leucine-rich repeat and fibronectin type-III domain-containing protein 5, SALM5FLJ30803
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human LRFN5. The immunogen is located within amino acids 650 - 700 of LRFN5. Amino Acid Squence: TNVESQNTNRNNSTAL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 145581
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.