Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRH-1/NR5A2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227196SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227196SS 0.025 mg
NBP227196 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227196SS Supplier Novus Biologicals Supplier No. NBP227196SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

LRH-1/NR5A2 Polyclonal specifically detects LRH-1/NR5A2 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Chinese Hamster, Equine, Guinea Pig, Opossum, Primate, Rabbit, Rhesus Macaque, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen LRH-1/NR5A2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-5ug/ml∼, Immunohistochemistry, Immunohistochemistry-Paraffin 5ug/ml, KnockDown Validated reported in scientific literature (PMID 31978970)
Formulation PBS with 0.05% Sodium Azide
Gene Alias Alpha-1-fetoprotein transcription factor, B1-binding factor, b1-binding factor, hepatocyte transcription factor which activates enhancer IIof hepatitis B virus, B1F, B1F2, fetoprotein-alpha 1 (AFP) transcription factor, FTFLRH1, FTZ-F1, FTZ-F1beta, HB1F, hB1F-2, hB1FCYP7A promoter-binding factor, Hepatocytic transcription factor, Liver receptor homolog 1, liver receptor homolog-1, LRH-1CPF, nuclear receptor NR5A2, nuclear receptor subfamily 5 group A member 2, nuclear receptor subfamily 5, group A, member 2
Gene Symbols NR5A2
Host Species Rabbit
Immunogen A portion of amino acids 140-190 from human NR5A2 was used as the immunogen for this antibody.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Chromatin Research, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2494
Test Specificity The amino acid sequence used as immunogen for this NR5A2 antibody is 100% homologous in human, mouse, rat, rhesus monkey, orangutan, chimpanzee, new world monkey, pig, panda, dog, Chinese hamster, horse, cow, african bush elephant, sheep, guinea pig, Carolina anole, zebrafish, chicken, great ape, mole rat, opossum, white-cheeked gibbon, platypus, rabbit, zebra finch, frog, and catfish.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Chinese Hamster, Equine, Guinea Pig, Opossum, Primate, Rabbit
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.