Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRH-1/NR5A2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$206.50 - $426.00

Specifications

Antigen LRH-1/NR5A2
Concentration 1.0 mg/ml
Dilution Western Blot 1-5ug/ml∼, Immunohistochemistry, Immunohistochemistry-Paraffin 5ug/ml, KnockDown Validated reported in scientific literature (PMID 31978970)
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227196SS
SDP
View Documents
Novus Biologicals
NBP227196SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP227196
SDP
View Documents
Novus Biologicals
NBP227196
0.1 mg
Each for $426.00
Only null left
Add to Cart
 
Description

Description

LRH-1/NR5A2 Polyclonal specifically detects LRH-1/NR5A2 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Chinese Hamster, Equine, Guinea Pig, Opossum, Primate, Rabbit, Rhesus Macaque, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications

Specifications

LRH-1/NR5A2
Western Blot 1-5ug/ml∼, Immunohistochemistry, Immunohistochemistry-Paraffin 5ug/ml, KnockDown Validated reported in scientific literature (PMID 31978970)
Polyclonal
Purified
RUO
Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Chinese Hamster, Equine, Guinea Pig, Opossum, Primate, Rabbit
Alpha-1-fetoprotein transcription factor, B1-binding factor, b1-binding factor, hepatocyte transcription factor which activates enhancer IIof hepatitis B virus, B1F, B1F2, fetoprotein-alpha 1 (AFP) transcription factor, FTFLRH1, FTZ-F1, FTZ-F1beta, HB1F, hB1F-2, hB1FCYP7A promoter-binding factor, Hepatocytic transcription factor, Liver receptor homolog 1, liver receptor homolog-1, LRH-1CPF, nuclear receptor NR5A2, nuclear receptor subfamily 5 group A member 2, nuclear receptor subfamily 5, group A, member 2
NR5A2
IgG
Protein A purified
The amino acid sequence used as immunogen for this NR5A2 antibody is 100% homologous in human, mouse, rat, rhesus monkey, orangutan, chimpanzee, new world monkey, pig, panda, dog, Chinese hamster, horse, cow, african bush elephant, sheep, guinea pig, Carolina anole, zebrafish, chicken, great ape, mole rat, opossum, white-cheeked gibbon, platypus, rabbit, zebra finch, frog, and catfish.
1.0 mg/ml
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown
Unconjugated
Rabbit
Chromatin Research, Neuroscience
PBS with 0.05% Sodium Azide
2494
A portion of amino acids 140-190 from human NR5A2 was used as the immunogen for this antibody.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.