Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRH-1/NR5A2 Antibody (OTI3H8), PerCP, Novus Biologicals™
SDP

Catalog No. NB288078 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288078 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288078 Supplier Novus Biologicals Supplier No. NBP272521PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LRH-1/NR5A2 Monoclonal antibody specifically detects LRH-1/NR5A2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen LRH-1/NR5A2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3H8
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias Alpha-1-fetoprotein transcription factor, B1-binding factor, b1-binding factor, hepatocyte transcription factor which activates enhancer IIof hepatitis B virus, B1F, B1F2, fetoprotein-alpha 1 (AFP) transcription factor, FTFLRH1, FTZ-F1, FTZ-F1beta, HB1F, hB1F-2, hB1FCYP7A promoter-binding factor, Hepatocytic transcription factor, Liver receptor homolog 1, liver receptor homolog-1, LRH-1CPF, nuclear receptor NR5A2, nuclear receptor subfamily 5 group A member 2, nuclear receptor subfamily 5, group A, member 2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-303 of human NR5A2(NP_995582) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2494
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.