Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRIG1 Antibody (789211), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB311223 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311223 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311223 Supplier Novus Biologicals Supplier No. FAB7498PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LRIG1 Monoclonal antibody specifically detects LRIG1 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen LRIG1
Applications Flow Cytometry
Classification Monoclonal
Clone 789211
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias D6Bwg0781e, Img, LIG-1
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human LRIG1, Ala35-Ser779, Accession # Q96JA1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 26018
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.