Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRIG1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP230926 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP230926 0.1 mL
NB397641 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP230926 Supplier Novus Biologicals Supplier No. NBP230926
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRIG1 Polyclonal specifically detects LRIG1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen LIG1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200, Knockdown Validated
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q96JA1
Gene Alias Leucine-Rich Repeat Protein LRIG1, Leucine-Rich Repeats And Immunoglobulin-Like Domains 1, Leucine-Rich Repeats And Immunoglobulin-Like Domains Protein 1, LIG-1, LRIG1, Ortholog Of Mouse Integral Membrane Glycoprotein LIG-1
Gene Symbols LIG1
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: TRKKSEEYSVTNTDETVVPPDVPSYLSSQGTLSDRQETVVRTEGGPQANGHIESNGVCPRDASHFPEPDTHSVACRQPKLCAGSAYHKEPWKAMEKAEGTPGPHKMEHGGRVVCSDCNTEVDCYS
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 26018
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.