Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRIG2 Antibody (990310), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB366973 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB366973 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB366973 Supplier Novus Biologicals Supplier No. FAB1941Y
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LRIG2 Monoclonal specifically detects LRIG2 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry.

Specifications

Antigen LRIG2
Applications CyTOF, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 990310
Conjugate DyLight 680
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias KIAA0806, leucine-rich repeats and immunoglobulin-like domains 2, leucine-rich repeats and immunoglobulin-like domains protein 2, LIG2
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human LRIG-2, Gly41-Thr805, Accession # O94898
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9860
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.