Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRIG3 Antibody (814834) [Janelia Fluor™ 646], Novus Biologicals™
SDP

Catalog No. NB372200 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB372200 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB372200 Supplier Novus Biologicals Supplier No. FAB3495J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LRIG3 Monoclonal antibody specifically detects LRIG3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen LRIG3
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 814834
Conjugate Janelia Fluor 646
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias FLJ90440, KIAA3016, leucine-rich repeats and immunoglobulin-like domains 3, leucine-rich repeats and immunoglobulin-like domains protein 3, LIG-3, LIG3FLJ26573, LIRG3
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LRIG3, Asp28-Thr807, Accession # Q6UXM1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121227
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.