Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRP-1 Antibody (A2Mr alpha-2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB100648080.025MG
Description
LRP-1 Monoclonal specifically detects LRP-1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Immunofluorescence.Specifications
| Low Density LRP | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot Non-reducing conditions, Flow Cytometry 1:50-1:100, ELISA Direct, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Q07954 | |
| LRP1 | |
| Purified alpha2 macroglobulin receptor | |
| 0.025 mg | |
| Primary | |
| Recognises an epitope in the extracellular region of the 500kD alpha chain. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Immunoprecipitation, ELISA, Immunohistochemistry, Western Blot, Flow Cytometry, Immunohistochemistry (Frozen) | |
| A2Mr alpha-2 | |
| Unconjugated | |
| Phosphate buffered saline with 0.09% Sodium Azide | |
| A2MRTGFBR5, Alpha-2-macroglobulin receptorMGC88725, APOER, Apolipoprotein E receptor, APR, CD91, CD91 antigen, EC 1.1.1.94, EC 3.4.21.9, FLJ16451, IGFBP3R, low density lipoprotein receptor-related protein 1, LRP, LRP-1, prolow-density lipoprotein receptor-related protein 1, TbetaR-V/LRP-1/IGFBP-3 receptor, type V tgf-beta receptor | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 4035 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction