Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRP-1 Antibody (A2Mr alpha-2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
$207.00 - $550.50
Specifications
| Antigen | Low Density LRP |
|---|---|
| Clone | A2Mr alpha-2 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot Non-reducing conditions, Flow Cytometry 1:50-1:100, ELISA Direct, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Immunoprecipitation, ELISA, Immunohistochemistry, Western Blot, Flow Cytometry, Immunohistochemistry (Frozen) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017648
![]() |
Novus Biologicals
NB100648080.025MG |
0.025 mg |
Each for $207.00
|
|
|||||
NB10064808
![]() |
Novus Biologicals
NB10064808 |
0.1 mg |
Each for $550.50
|
|
|||||
Description
LRP-1 Monoclonal specifically detects LRP-1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Immunofluorescence.Specifications
| Low Density LRP | |
| 1.0 mg/mL | |
| Immunoprecipitation, ELISA, Immunohistochemistry, Western Blot, Flow Cytometry, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Q07954 | |
| 4035 | |
| Purified alpha2 macroglobulin receptor | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| A2Mr alpha-2 | |
| Western Blot Non-reducing conditions, Flow Cytometry 1:50-1:100, ELISA Direct, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| A2MRTGFBR5, Alpha-2-macroglobulin receptorMGC88725, APOER, Apolipoprotein E receptor, APR, CD91, CD91 antigen, EC 1.1.1.94, EC 3.4.21.9, FLJ16451, IGFBP3R, low density lipoprotein receptor-related protein 1, LRP, LRP-1, prolow-density lipoprotein receptor-related protein 1, TbetaR-V/LRP-1/IGFBP-3 receptor, type V tgf-beta receptor | |
| LRP1 | |
| IgG1 | |
| Protein G purified | |
| Recognises an epitope in the extracellular region of the 500kD alpha chain. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title