Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRP-1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP262752 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP262752 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP262752 Supplier Novus Biologicals Supplier No. NBP262752

Rabbit Polyclonal Antibody

LRP-1 Polyclonal specifically detects LRP-1 in Rat samples. It is validated for Western Blot.

Specifications

Antigen LRP-1
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide
Gene Accession No. Q07954
Gene Alias A2MR, Alpha-2-Macroglobulin Receptor, APOER, Apolipoprotein E receptor, APR, CD91, CD91 antigen, EC 1.1.1.94, EC 3.4.21.9, IGFBP3R, low density lipoprotein receptor-related protein 1, LRP, LRP-1, LRP1A, prolow-density lipoprotein receptor-related protein 1, TbetaR-V/LRP-1/IGFBP-3 receptor, TGFBR5, type V tgf-beta receptor
Gene Symbols LRP1
Host Species Rabbit
Immunogen Synthetic peptide from the mid-protein of Human Low-density lipoprotein receptor-related protein 1 (aa. 2880-2980)
Purification Method Peptide affinity purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 4035
Test Specificity Detects 80 kDa, 50 kDa. Band at 80 kDa is a membrane spanning subunit, 50 kDa may be a degradation product.
Target Species Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.