Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRP-1 Antibody, Novus Biologicals™
SDP

Catalog No. NB413791 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB413791 0.025 mg
NBP262753 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB413791 Supplier Novus Biologicals Supplier No. NBP2627530.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRP-1 Polyclonal specifically detects LRP-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen LRP-1
Applications Flow Cytometry, Immunohistochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:50
Formulation PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide
Gene Alias A2MR, Alpha-2-Macroglobulin Receptor, APOER, Apolipoprotein E receptor, APR, CD91, CD91 antigen, EC 1.1.1.94, EC 3.4.21.9, IGFBP3R, low density lipoprotein receptor-related protein 1, LRP, LRP-1, LRP1A, prolow-density lipoprotein receptor-related protein 1, TbetaR-V/LRP-1/IGFBP-3 receptor, TGFBR5, type V tgf-beta receptor
Gene Symbols LRP1
Host Species Rabbit
Immunogen Synthetic peptide from the N-terminal of Human Low-density lipoprotein receptor-related protein 1 (aa. 160-260)
Purification Method Affinity Purified
Quantity 0.025 mg
Primary or Secondary Primary
Gene ID (Entrez) 4035
Test Specificity Detects 80 kDa. Band at 80 kDa is a membrane spanning subunit.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.