Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRP-11 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB065000 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB065000 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB065000 Supplier Novus Biologicals Supplier No. NBP300238DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRP-11 Polyclonal specifically detects LRP-11 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen LRP-11
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias bA350J20.3, FLJ14735, low density lipoprotein receptor-related protein 11, low-density lipoprotein receptor-related protein 11, LRP-11, MANSC3, MGC39092
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant LRP-11 (Q86VZ4-1; Met1-Gly450).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84918
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.