Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRP-4 Antibody (741704) [DyLight 350], Novus Biologicals™
SDP

Catalog No. NB377276 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB377276 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB377276 Supplier Novus Biologicals Supplier No. FAB5948D

Mouse Monoclonal Antibody

LRP-4 Monoclonal antibody specifically detects LRP-4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry

Specifications

Antigen LRP-4
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 741704
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias CLSS, KIAA0816, low density lipoprotein receptor-related protein 4, low-density lipoprotein receptor-related protein 4, LRP-4, MEGF7LRP10, Multiple epidermal growth factor-like domains 7
Host Species Mouse
Immunogen E. coli-derived recombinant human LRP-4, Gly18-Ser344, Accession # O75096
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4038
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.