Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRP-4 Antibody (S207-27), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB222416700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB222416700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB222416700 Supplier Novus Biologicals Supplier No. NBP222416AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LRP-4 Monoclonal specifically detects LRP-4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen LRP-4
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S207-27
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CLSS, KIAA0816, low density lipoprotein receptor-related protein 4, low-density lipoprotein receptor-related protein 4, LRP-4, MEGF7LRP10, Multiple epidermal growth factor-like domains 7
Gene Symbols LRP4
Host Species Mouse
Immunogen Fusion protein amino acids 26- 350 (extracellular N-terminus) of mouse LTP4
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4038
Test Specificity Detects the extracellular domain of LRP4 approx 215kDa. Degradation product can be seen at approx 150kDa.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.