Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRP-6 Antibody (255302), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB349888 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB349888 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB349888 Supplier Novus Biologicals Supplier No. FAB1505MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LRP-6 Monoclonal antibody specifically detects LRP-6 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen LRP-6
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 255302
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias ADCAD2, FLJ90062, FLJ90421, low density lipoprotein receptor-related protein 6, low-density lipoprotein receptor-related protein 6, LRP-6
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LRP-6, Ala20-Pro1368, Accession # O75581
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4040
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.