Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC15 Antibody (2944A), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB297421 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297421 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB297421 Supplier Novus Biologicals Supplier No. FAB11324W
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

LRRC15 Monoclonal antibody specifically detects LRRC15 in Human samples. It is validated for Western Blot, Western Blot, Flow Cytometry

Specifications

Antigen LRRC15
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 2944A
Conjugate DyLight 650
Dilution Western Blot, Simple Western, Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias hLib, leucine rich repeat containing 15
Host Species Rabbit
Immunogen Chinese Hamster Ovary cell line, CHO-derived human LRRC15, Met1-Gly538, Accession # Q8TF66
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 131578
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.