Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC15 Antibody (2944A), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB314867 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB314867 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB314867 Supplier Novus Biologicals Supplier No. FAB11324PEATT594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

LRRC15 Monoclonal specifically detects LRRC15 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen LRRC15
Applications Flow Cytometry
Classification Monoclonal
Clone 2944A
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias hLib, leucine rich repeat containing 15
Host Species Rabbit
Immunogen Chinese Hamster Ovary cell line, CHO-derived human LRRC15, Met1-Gly538, Accession # Q8TF66
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 131578
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.