Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC19 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB164269 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164269 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB164269 Supplier Novus Biologicals Supplier No. NBP281732F
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LRRC19 Polyclonal antibody specifically detects LRRC19 in Human, Mouse samples. It is validated for ELISA, Immunofluorescence

Specifications

Antigen LRRC19
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias leucine rich repeat containing 19
Host Species Rabbit
Immunogen LRRC19 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human LRRC19. The immunogen is located within amino acids 290 - 340 of LRRC19. Amino Acid Squence: YNHHRLEEHEAETYEDG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 64922
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.