Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC25 Antibody (OTI1D9), PerCP, Novus Biologicals™
SDP

Catalog No. NB272523PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB272523PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB272523PCP Supplier Novus Biologicals Supplier No. NBP272523PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LRRC25 Monoclonal specifically detects LRRC25 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen LRRC25
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1D9
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias leucine rich repeat containing 25, leucine-rich repeat-containing protein 25, MAPAFLJ38116, monocyte and plasmacytoid activated molecule, Monocyte and plasmacytoid-activated protein
Gene Symbols LRRC25
Host Species Mouse
Immunogen Full length recombinant protein of human LRRC25 (NP_660299) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 126364.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.