Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC32/GARP Antibody (725226) [DyLight 405], Novus Biologicals™
SDP

Catalog No. NB376862 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB376862 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB376862 Supplier Novus Biologicals Supplier No. FAB62291E
Add to Cart
Add to Cart

Rat Monoclonal Antibody

LRRC32/GARP Monoclonal antibody specifically detects LRRC32/GARP in Mouse samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen LRRC32/GARP
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 725226
Conjugate DyLight 405
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias D11S833Eleucine-rich repeat-containing protein 32, GARPGarpin, Glycoprotein A repetitions predominantgarpin, leucine rich repeat containing 32
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse LRRC32/GARP, Ile18-Asn628, Accession # NP_001106850
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2615
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.