Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC32/GARP Antibody (855151), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB377075 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB377075 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB377075 Supplier Novus Biologicals Supplier No. FAB6055I
Add to Cart
Add to Cart

Rat Monoclonal Antibody

LRRC32/GARP Monoclonal specifically detects LRRC32/GARP in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen LRRC32/GARP
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 855151
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias D11S833Eleucine-rich repeat-containing protein 32, GARPGarpin, Glycoprotein A repetitions predominantgarpin, leucine rich repeat containing 32
Host Species Rat
Immunogen recombinant human LRRC32/GARP, Accession # Q14392
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2615
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.