Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC32/GARP Antibody, Novus Biologicals™
SDP

Catalog No. NB392923 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392923 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB392923 Supplier Novus Biologicals Supplier No. NBP26874025UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRRC32/GARP Polyclonal antibody specifically detects LRRC32/GARP in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/ Immunofluorescence

Specifications

Antigen LRRC32/GARP
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias D11S833Eleucine-rich repeat-containing protein 32, GARPGarpin, Glycoprotein A repetitions predominantgarpin, leucine rich repeat containing 32
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: GNPLSCCGNGWLAAQLHQGRVDVDATQDLICRFSSQEEVSLSHVRPEDCEK
Purification Method Protein A purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2615
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.