Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC35 Antibody (1B10), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB160837 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB160837 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB160837 Supplier Novus Biologicals Supplier No. NBP259417AF405
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LRRC35 Monoclonal antibody specifically detects LRRC35 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen LRRC35
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 1B10
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias tubulin-specific chaperone cofactor E-like protein, El, LRRC35, tubulin folding cofactor E-like
Host Species Mouse
Immunogen Recombinant human LRRC35 (1-424aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219899
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.