Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC3B/LRP15 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB293684 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB293684 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB293684 Supplier Novus Biologicals Supplier No. NBP326069R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRRC3B/LRP15 Polyclonal antibody specifically detects LRRC3B/LRP15 in Human samples. It is validated for ELISA

Specifications

Antigen LRRC3B/LRP15
Applications ELISA
Classification Polyclonal
Conjugate DyLight 550
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias leucine rich repeat containing 3B, leucine-rich repeat-containing protein 3B, LRP15Leucine-rich repeat protein LRP15, MGC102927
Host Species Rabbit
Immunogen Rabbits immunized with purified, recombinant Human LRRC3B/LRP15 (Accession#: NP_443185.1, Met1-Tyr204)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116135
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.