Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC48 Antibody, Novus Biologicals™
SDP

Catalog No. NB404675 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB404675 25 μL
NBP238277 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB404675 Supplier Novus Biologicals Supplier No. NBP23827725UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRRC48 Polyclonal specifically detects LRRC48 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen LRRC48
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q9H069
Gene Alias DKFZP586M1120, leucine rich repeat containing 48, leucine-rich repeat-containing protein 48
Gene Symbols LRRC48
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: GVGELLETYKDKFVIICVNIFEYGLKQQEKRKTELDTFSECVREAIQENQEQGKRKIAKFEEKHLSSLSAIREELELPNIEKMILECSADISELFDALMTLEMQLVEQ
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83450
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.